Anti-GAPDH Mouse Monoclonal Antibody [clone: 13k57]
USBI143184
- Antibody type:Primary
- Antigen name:Glyceraldehyde-3-phosphate dehydrogenase
- Antigen symbol:GAPDH
- Clonality:Monoclonal
- Clone:13k57
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM NaCl, 0,2% sodium azide, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
- Recommended dilution: Western Blot: 1:5000
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone: 13k57
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey