Drukuj

Anti-B. anthracis SA Mouse Monoclonal Antibody [clone: 7C11]

Dostawca: United States Biological
USBIB0003-05G10EA 3560 PLN
USBIB0003-05G10
Anti-B. anthracis SA Mouse Monoclonal Antibody [clone: 7C11]
Przeciwciała
Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: B. anthracis SA (Bacillus anthracis spore antigen)
Clonality: monoclonal
Clone: 7C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR