Drukuj

Anti-Bacillus anthracis Spore Antigen (Anthrax) (Biotin) [clone: 3G302A]Anti-Bacillus anthracis Spore Antigen (Anthrax) (Biotin)

Dostawca: United States Biological
USBIB0003-05G31-25EA 2230 PLN
USBIB0003-05G31-25
Anti-Bacillus anthracis Spore Antigen (Anthrax) (Biotin) [clone: 3G302A]Anti-Bacillus anthracis Spore Antigen (Anthrax) (Biotin)
Przeciwciała
Anthrax toxin plays a major role in the pathogenesis of anthrax. Virulent strains of Bacillus anthracis produce a complex exotoxin with 3 distinct antigenic components: the edema factor (EF), the protective antigen or binding domain (PA), and the lethal factor (LF). Supplied as a liquid in PBS, pH 7.2. Labeled with Biotin.

Type: Primary
Antigen: B. anthracis SA (Bacillus anthracis spore antigen)
Clonality: monoclonal
Clone: 3G302A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR