Drukuj

Anti-GITRL Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIG2035-14A-50EA 1630 PLN
USBIG2035-14A-50
Anti-GITRL Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 0.5µg/ml

Type: Primary
Antigen: GITRL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR