Drukuj

Anti-GITRL Rat Polyclonal Antibody

Dostawca: United States Biological
USBIG2035-15EA 2760 PLN
USBIG2035-15
Anti-GITRL Rat Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: GITRL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR