Drukuj

Anti-POU6F2 Goat Polyclonal Antibody

Dostawca: United States Biological
USBIP5250-52EA 2770 PLN
USBIP5250-52
Anti-POU6F2 Goat Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: POU6F2 (POU class 6 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR