Drukuj

Anti-POTED Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dostawca: United States Biological
USBIP5280-01-FITCEA 4280 PLN
USBIP5280-01-FITC
Anti-POTED Rabbit Polyclonal Antibody (FITC (Fluorescein))
Przeciwciała
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: POTED (POTE ankyrin domain family, member D)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR