Chcesz zalogować się jeszcze raz?
Anti-PPARG Rabbit monoclonal antibody unconjugated [clone: 5H57]
Dostawca: US Biological
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: monoclonal
Clone: 5H57
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
ZAMÓW
Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µl 100 µl USBIP5505-01P
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol PPARG
Antigen name peroxisome proliferator-activated receptor gamma
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 5H57
Host Rabbit
Isotype IgG
Western blot Yes
Modification phospho-Ser82
Purification purified by Protein A chromatography
Form liquid
Storage buffer 0,2M Tris-glycine, pH 7,2, 0,15M sodium chloride and 0,05% sodium azide 30% glycerol.
Temperatura przechowywania Cool
Shipping temperature Ice
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość