Drukuj

Anti-PPARG Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP5505-05BEA 3250 PLN
USBIP5505-05B
Anti-PPARG Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR