Drukuj

Anti-PPARA Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP5505-07EA 4230 PLN
USBIP5505-07
Anti-PPARA Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:8000-1:32000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR