Drukuj
Anti-PPARA Rabbit Polyclonal Antibody
Dostawca: United States Biological
USBIP5505-07C1EA 3490 PLN
USBIP5505-07C1
Anti-PPARA Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:8000-1:32000; Western Blot: 1:500-1:2000; Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse