Drukuj

Anti-PPARA Mouse monoclonal antibody [clone: 8J138]

USBIP5505-07EEA 2120 PLN
USBIP5505-07E
Anti-PPARA Mouse monoclonal antibody [clone: 8J138]
Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: monoclonal
Clone: 8J138
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR