Drukuj

Anti-PPARD Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP5505-08AEA 2820 PLN
USBIP5505-08A
Anti-PPARD Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:2000-1:8000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: PPARD (peroxisome proliferator-activated receptor delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR