Drukuj
Anti-PPARD Rabbit Polyclonal Antibody
Dostawca: United States Biological
USBIP5505-08EEA 2850 PLN
USBIP5505-08E
Anti-PPARD Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: PPARD (peroxisome proliferator-activated receptor delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human