Drukuj
Anti-PPARg, Coactivator 1 alpha Mouse Monoclonal Antibody [clone: 8C270]
Dostawca: United States Biological
USBIP5505-55EA 2040 PLN
USBIP5505-55
Anti-PPARg, Coactivator 1 alpha Mouse Monoclonal Antibody [clone: 8C270]
Przeciwciała
Type: Primary
Antigen: PPARg, Coactivator 1 alpha
Clonality: monoclonal
Clone: 8C270
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human