Drukuj

Anti-PPNK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: United States Biological
USBIP5510-30A-APEA 4280 PLN
USBIP5510-30A-AP
Anti-PPNK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: PPNK
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR