Drukuj
Anti-PRMT2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dostawca: United States Biological
USBIP9004-10A-HRPEA 4280 PLN
USBIP9004-10A-HRP
Anti-PRMT2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: PRMT2 (protein arginine methyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human