Drukuj

Anti-PRMT2 Mouse monoclonal antibody [clone: 8C271]

USBIP9004-10BEA 2120 PLN
USBIP9004-10B
Anti-PRMT2 Mouse monoclonal antibody [clone: 8C271]
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: PRMT2 (protein arginine methyltransferase 2)
Clonality: monoclonal
Clone: 8C271
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR