Drukuj

Anti-PRMT2 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP9004-10CEA 2950 PLN
USBIP9004-10C
Anti-PRMT2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: PRMT2 (protein arginine methyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR