Drukuj

Anti-PRMT3 Rabbit Polyclonal Antibody (Biotin)

Dostawca: United States Biological
USBIP9004-17A-BIOTEA 4280 PLN
USBIP9004-17A-BIOT
Anti-PRMT3 Rabbit Polyclonal Antibody (Biotin)
Przeciwciała
Type: Primary
Antigen: PRMT3 (protein arginine methyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR