Drukuj

Anti-PRMT3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dostawca: United States Biological
USBIP9004-17B-HRPEA 4280 PLN
USBIP9004-17B-HRP
Anti-PRMT3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PRMT3 (protein arginine methyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR