Drukuj
Anti-PRMT5 Rabbit Polyclonal Antibody (Biotin)
Dostawca: United States Biological
USBIP9004-25B-BIOTEA 4280 PLN
USBIP9004-25B-BIOT
Anti-PRMT5 Rabbit Polyclonal Antibody (Biotin)
Przeciwciała
Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human