Drukuj
Anti-PRMT5 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Dostawca: United States Biological
USBIP9004-25B-FITCEA 4280 PLN
USBIP9004-25B-FITC
Anti-PRMT5 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human