Drukuj

Anti-PRMT5 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP9004-26EA 4670 PLN
USBIP9004-26
Anti-PRMT5 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR