Drukuj

Anti-PRMT6 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP9004-30GEA 3180 PLN
USBIP9004-30G
Anti-PRMT6 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-100;

Type: Primary
Antigen: PRMT6 (protein arginine methyltransferase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR