Drukuj

Anti-PRMT7 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP9004-35EEA 2260 PLN
USBIP9004-35E
Anti-PRMT7 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: PRMT7 (protein arginine methyltransferase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR