Anti-SLC1A5 Rabbit Polyclonal Antibody
USBIS5329-20
- Antibody type:Primary
- Antigen name:solute carrier family 1 (neutral amino acid transporter), member 5
- Antigen symbol:Solute carrier family 1, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:193 to 222
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Recommended dilution: Western Blot: 1:50-1:100
Type: Primary
Antigen: SLC1A5 (solute carrier family 1 (neutral amino acid transporter), member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human