Drukuj

Anti-WNT3A Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIW3011-02CEA 2780 PLN
USBIW3011-02C
Anti-WNT3A Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: WNT3A (wingless-type MMTV integration site family, member 3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR