Drukuj

Anti-WNT4 Goat Polyclonal Antibody

Dostawca: United States Biological
USBIW3021-01EA 2770 PLN
USBIW3021-01
Anti-WNT4 Goat Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR