Drukuj
Anti-WNT4 Rabbit Polyclonal Antibody
Dostawca: United States Biological
USBIW3021-01AEA 2950 PLN
USBIW3021-01A
Anti-WNT4 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human