Drukuj

Anti-WNT4 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIW3021-01BEA 2950 PLN
USBIW3021-01B
Anti-WNT4 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR