Drukuj

Anti-WNT5A Mouse Monoclonal Antibody [clone: 6F2]

Dostawca: United States Biological
USBIW3028-01CEA 3180 PLN
USBIW3028-01C
Anti-WNT5A Mouse Monoclonal Antibody [clone: 6F2]
Przeciwciała
Recommended dilution: Immunofluorescence: 1:200-1:1000; ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:400

Type: Primary
Antigen: WNT5A (wingless-type MMTV integration site family, member 5A)
Clonality: monoclonal
Clone: 6F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR