Drukuj

Anti-WNT5A Rabbit polyclonal antibody

USBIW3028-01VEA 1440 PLN
USBIW3028-01V
Anti-WNT5A Rabbit polyclonal antibody
Recommended dilution: ELISA: 1:5000-1:20000

Recommended dilution: ELISA: 1:5000-1:20000

Type: Primary
Antigen: WNT5A (wingless-type MMTV integration site family, member 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR