Drukuj

Anti-WNT2B Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIW3035-40BEA 3540 PLN
USBIW3035-40B
Anti-WNT2B Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: WNT2B (wingless-type MMTV integration site family, member 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR