Drukuj

Anti-WNT3 Goat Polyclonal Antibody

Dostawca: United States Biological
USBIW3035-55AEA 3180 PLN
USBIW3035-55A
Anti-WNT3 Goat Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:2000; Western Blot: 1:2000; Immunochemistry: 3.75 ?g/ml

Type: Primary
Antigen: WNT3 (wingless-type MMTV integration site family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR