Drukuj

Anti-WNT5B Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dostawca: United States Biological
USBIW3035-92D-PEEA 4280 PLN
USBIW3035-92D-PE
Anti-WNT5B Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: WNT5B (wingless-type MMTV integration site family, member 5B)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR