Drukuj

Anti-WNT6 Sheep Polyclonal Antibody

Dostawca: United States Biological
USBIW3036-01EA 3980 PLN
USBIW3036-01
Anti-WNT6 Sheep Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 0.1-0.2?g/ml; Immunochemistry: 5-15 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: WNT6 (wingless-type MMTV integration site family, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR