Drukuj

Anti-WNT6 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIW3036-02EA 2260 PLN
USBIW3036-02
Anti-WNT6 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: WNT6 (wingless-type MMTV integration site family, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR