Chcesz zalogować się jeszcze raz?
Anti-WNT8B Rat monoclonal antibody unconjugated [clone: 7H100]
Dostawca: US Biological
Recommended dilution: Immunochemistry: 10 μg/ml
Type: Primary
Antigen: WNT8B (wingless-type MMTV integration site family, member 8B)
Clonality: monoclonal
Clone: 7H100
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
ZAMÓW
Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µg 100 µG USBIW3037-10
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol WNT8B
Antigen name wingless-type MMTV integration site family, member 8B
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 7H100
Host Rat
Isotype IgG2a
Western blot Yes
ELISA Yes
Stężenie ~0,5 mg/ml
Purification purified by Protein G chromatography
Form lyophilized powder
Storage buffer PBS, 5% trehalose.
Temperatura przechowywania Cool
Shipping temperature Ice
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość