Drukuj

Anti-WNT8B Rabbit polyclonal antibody

USBIW3037-12EA 2190 PLN
USBIW3037-12
Anti-WNT8B Rabbit polyclonal antibody
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1:5000-1:20000

Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1:5000-1:20000

Type: Primary
Antigen: WNT8B (wingless-type MMTV integration site family, member 8B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR