Drukuj
Zatyczki do uszu E-A-R™ Ultrafit™ 20
Dostawca: 3M
E-A-R™ UltraFit™
MMMAUF01012EA 1200 PLN
MMMAUF01012
Zatyczki do uszu E-A-R™ Ultrafit™ 20
Zatyczki do uszu Zatyczki do uszu wielokrotnego użytku
These pre-moulded earplugs are suitable for moderate noise levels (83 to 93 dB(A)). Correctly matched to the noise level they allow conversation and background music to be heard whilst helping to protect the user from hearing loss.
  • Moisture resistant and compatible with other PPE
  • No need for any roll down
  • Triple flanged tip, pistol grip stem and angled shape for easy insertion
  • Lightweight design (185 g)
  • Washable and reusable
SNR: 20 dB
Certyfikaty: EN 352-2.
Uwaga: * One size fits all.