Anaerobowy system inkubacji, naczynia beztlenowe
Supplier: THERMO FISHER DIAGNOSTICS
Certificates
The jar is designed for use with AnaeroGen/CampyGen sachet and will hold up to 12 plates.
- Both jar and lid are made of robust materials to ensure protection of operator and equipment
- No catalyst required
- A carrying handle for the safe transportation of the jar from bench to incubator
Product Information
- Catalog #Supplier #OpisIlośćAvailabilityPrice
- OXOIAN0020DAN0020DAnaerogen compact101 * 10 SZT227 złSupplier Article Number: AN0020DLocal Article Number: OXOIAN0020D
Accessories
Specifications
Environmentally Preferable - OXOICN0025ACN0025ACampygen 2,5 l101 * 10 SZT281 złSupplier Article Number: CN0025ALocal Article Number: OXOICN0025A
Accessories
Specifications
Environmentally Preferable - OXOICN0035ACN0035ACampygen 3,5 l101 * 10 SZT320 złSupplier Article Number: CN0035ALocal Article Number: OXOICN0035A
Accessories
Specifications
Environmentally Preferable - OXOIAG0025AAG0025AAnaerojar 2,5 l11 * 1 SZT2 020 złSupplier Article Number: AG0025ALocal Article Number: OXOIAG0025A
Accessories
Specifications
Environmentally Preferable - OXOICN0020CCN0020CCampygen Compact201 * 20 SZT356 złSupplier Article Number: CN0020CLocal Article Number: OXOICN0020C
Accessories
Specifications
Environmentally Preferable