Drukuj
Medium do hodowli, gotowe do użycia, w probówkach
Dostawca: Sifin Diagnostics GmbH
SIFITN1517EA 301 PLN
SIFITN1517 SIFITN1516 SIFITN1513 SIFITN1515 715-9694
Medium do hodowli, gotowe do użycia, w probówkach
Media do mikrobiologii Podłoża gotowe do użycia Podłoża gotowe do użycia w probówkach
Manufacturer for more than 250 different culture media and supplements in order to cultivate the microorganisms or specifically promote the growth of particularly fastidious species. Thus, you are optimally equipped for the analysis of veterinary or human samples.
  • Special media for food, water, pharmaceutical products complete the portfolio.