Drukuj
Lepkościomierze Ubbelohde
Dostawca: Lauda
LAUDEGV700EA 0 PLN
LAUDEGV700 LAUDEGV703 LAUDEGV717 LAUDEGV706 LAUDEGV707 LAUDEGV701 LAUDEGV702 LAUDEGV705 LAUDEGV699 LAUDEGV716 LAUDEGV710 LAUDEGV712 LAUDEGV697 LAUDEGV715 LAUDEGV704 LAUDEGV708
Lepkościomierze Ubbelohde
Ubbelohde viscometers calibrated for automatic and stopwatch measurements or uncalibrated.
  • No thread to facilitate the maintenance and cleaning
  • Corrosion resistance to strong acids and aggressive chemicals
  • Integrated frit to ensure filtration when the sample dissolves in the viscometer
Certyfikaty: ISO 3105, DIN 51562, BS 188, NFT 60-100 − Also available in ASTM version.