The innovative VWR Tablet System expands the capability of selected VWR balances and moisture analysers by increasing the functionality and applications available, using dedicated software and the now familiar 7 inch tablet touch screen technology. The small Tablet System provides an economical, powerful and space saving alternative to external computer control or use of more elaborate balances with larger displays.
- Expand the capability and versatility of existing VWR balances and moisture analysers with easy to use touch screen technology
- Connect to instrument and the tablet automatically recognises the instrument (no need to configure)
- See real time graph of drying process for moisture analysers
- Weighing/drying data and configuration of operations are shown on a large 7 inch touch screen display
- Screenshots can be easily taken and sent by email for fast comunication of work data to colleagues
- The tablet can retrieve data and information relevant to measurements/ongoing studies via the Internet - no need to move from another working location
- Instructions, in different languages, for various functions are stored in the tablet and accessible directly from the interface easily and quickly
VWR balances and moisture analysers compatible with the Tablet System:
LA analytical balances, LCD display; from 0,0001 to 0,01 g resolution (but not LAG balances)
LPW precision balances, LCD display; 0,001 g resolution (but not LPG, LPCG or LPWG balances)
LP precision balances, LCD display; from 0,001 to 0,1 g resolution (including 611-2312, 611-2313, 611-2314, 611-2315, 611-2316)
SE balances 611-2602 and 611-2604
IS series high capacity balances
MB 64 and MB 160 moisture analysers
Please note that the VWR Tablet System is not accredited/suitable for use with verified or EC-type approved balances.
The tablet's Bluetooth® connections and the Tablet System can be easily updated and applications upgraded via the Internet, without the sometimes necessary purchase of a new balance or moisture analyser. New applications will be developed in future which will expand the capabilities of selected instruments.
Dodatkowe informacje: The Tablet System includes a 7 inch tablet multi-touch screen, with Android operating system; Tab-Box connecting interface to connect the balance or moisture analyser to the tablet; pre-installed application with functions as described below; tilting tablet support.
Uwaga: Tablet Systems are not available for LAG and LPxG models (large graphic display analytical and precision balances) and touch screen versions of the moisture balance (MBT 160).