Znaki bezpieczeństwa
Supplier: Brady
Certificates
About this item
A comprehensive range of Emergency Escape and First Aid signs which help comply with current legislation.
- Laminated polyester
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Good weatherability and UV resistance
- Excellent adhesion to low surface energy surfaces
- Available in different materials and sizes
Product Information
- Catalog #Supplier #OpisMateriałWymiaryIlośćAvailabilityPrice
- BRDY814605814605First aid (E003)PP200×200 mm11 * 1 SZT58,70 złSpecifications:
Specifications
Environmentally Preferable - 116-0564814411Direction arrow (90° increments), safe condition (A090)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0565814414Direction arrow (90° increments), safe condition (A090)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - BRDY814604814604First aid (E003)PP148×148 mm11 * 1 SZT51,70 złSpecifications:
Specifications
Environmentally Preferable - 116-0562814438Emergency exit right (E002)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0563814441Emergency exit right (E002)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0560814597First aid (E003)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0561814600First aid (E003)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0573815713Safety shower (E012)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0576814915Evacuation assembly point (E007)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0577814918Evacuation assembly point (E007)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0574815551Eyewash station (E011)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0575815554Eyewash station (E011)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0580814756Emergency telephone (E004)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0568816028Emergency window with escape ladder (E016)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0581814759Emergency telephone (E004)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0566814384Direction arrow 45° (90° increments), safe condition (A045)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0578814265Emergency exit left (E001)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0567814387Direction arrow 45° (90° increments), safe condition (A045)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0579814268Emergency exit left (E001)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - 116-0572815710Safety shower (E012)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - 116-0570816187Rescue window (E017)PE200×200 mm11 * 1 SZT27,40 złSpecifications:
Specifications
Environmentally Preferable - BRDY814446814446Emergency exit right (E002)PP200×200 mm11 * 1 SZT58,70 złSpecifications:
Specifications
Environmentally Preferable - BRDY814392814392Direction arrow 45° (90° increments), safe condition (A045)PP200×200 mm11 * 1 SZT58,70 złSpecifications:
Specifications
Environmentally Preferable - BRDY814419814419Direction arrow (90° increments), safe condition (A090)PP200×200 mm11 * 1 SZT58,70 złSpecifications:
Specifications
Environmentally Preferable - 116-0569816031Emergency window with escape ladder (E016)PE315×315 mm11 * 1 SZT70,20 złSpecifications:
Specifications
Environmentally Preferable - BRDY814273814273Emergency exit left (E001)PP200×200 mm11 * 1 SZT58,70 złSpecifications:
Specifications
Environmentally Preferable