Order Entry
Headspace vials, Supelco®
Headspace vials, Supelco®
 :  Merck
Headspace vials, Supelco®
 :  Merck
  •  
    Capacity
    Ø×H
    Material
    Colour
    Cap included
  • Headspace vials, 12,5 mm
    Headspace vials, 12,5 mm
    SIALSU860029
    SU860029
    10,00 ml
    22,5×46 mm
    Glass
    Clear
    No
    1 * 1 Szt.
    1 * 1 Szt. - 162 zł
    162 zł
    1 * 1 Szt.
       162 zł
     : SU860029
     : SIALSU860029
    Specifications
    Environmentally Preferable
  • Headspace vials
    Headspace vials
    SIALSU860134
    SU860134
    50,00 ml
    31×101 mm
    Glass
    Clear
    No
    1 * 1.000 SZT
    1 * 1.000 SZT
     : SU860134
     : SIALSU860134
    Specifications
    Environmentally Preferable