Order Entry
Bottles, low particle
undefined
Bottles, low particle
Specifications
  • Catalog No:
  • EPSC120-04A/LP
  • EPSC113-500A/LP
  • EPSC120-02A/LP
  • EPSC114-125A/LP
  • EPSC114-060A/LP
  • EPSC114-250A/LP
  • EPSC114-125CT/LP
  • EPSC114-250C/LP
  • EPSC110-80A/LP
  • EPSC112-01A/LP
  • EPSC140-40C/LP
  • EPSC111-04A/LP
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Amber
  •  
    Capacity
    Ext.Ø
    Height
    Colour
    Cap Size
  • Packaging bottle with PTFE lined closure
    Packaging bottle with PTFE lined closure
    EPSC120-02A/LP
    120-02A/LP
    57 ml
    44,45 mm
    76,2 mm
    Amber
    33 - 400 mm
    1 * 24 SZT
    1 * 24 SZT - 2 570 zł
    2 570 zł
    1 * 24 SZT
       2 570 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Packaging bottle with PTFE lined closure
    Packaging bottle with PTFE lined closure
    EPSC120-04A/LP
    120-04A/LP
    125 ml
    50,8 mm
    95,25 mm
    Amber
    38 - 400 mm
    1 * 12 SZT
    1 * 12 SZT - 1 930 zł
    1 930 zł
    1 * 12 SZT
       1 930 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Boston round bottle with open-top septa closure with PTFE/silicone liner
    Boston round bottle with open-top septa closure with PTFE/silicone liner
    EPSC114-060A/LP
    114-060A/LP
    60 ml
    47,63 mm
    111,13 mm
    Amber
    20 - 400 mm
    1 * 24 SZT
    1 * 24 SZT - 2 380 zł
    2 380 zł
    1 * 24 SZT
       2 380 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Boston round bottle with closed top with screw cap with PTFE/silicone liner
    Boston round bottle with closed top with screw cap with PTFE/silicone liner
    EPSC114-125A/LP
    114-125A/LP
    125 ml
    47,63 mm
    111,13 mm
    Amber
    24 - 414 mm
    1 * 12 SZT
    1 * 12 SZT - 2 350 zł
    2 350 zł
    1 * 12 SZT
       2 350 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Boston round bottle with closed top with screw cap with PTFE/silicone liner
    Boston round bottle with closed top with screw cap with PTFE/silicone liner
    EPSC114-250A/LP
    114-250A/LP
    250 ml
    60,33 mm
    136,53 mm
    Amber
    24 - 414 mm
    1 * 12 SZT
    1 * 12 SZT - 2 390 zł
    2 390 zł
    1 * 12 SZT
       2 390 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Boston round bottle with closed top with screw cap with PTFE liner
    Boston round bottle with closed top with screw cap with PTFE liner
    EPSC113-500A/LP
    113-500A/LP
    500 ml
    76,2 mm
    168,28 mm
    Amber
    28 - 400 mm
    1 * 12 SZT
    1 * 12 SZT - 2 780 zł
    2 780 zł
    1 * 12 SZT
       2 780 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Flurinated glass jug with screw cap with PTFE/silicone liner
    Flurinated glass jug with screw cap with PTFE/silicone liner
    EPSC110-80A/LP
    110-80A/LP
    2500 ml
    133,4 mm
    305 mm
    Amber
    38 - 430 mm
    1 * 6 SZT
    1 * 6 SZT - 1 970 zł
    1 970 zł
    1 * 6 SZT
       1 970 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Glass bottle with screw cap with PTFE/silicone liner
    Glass bottle with screw cap with PTFE/silicone liner
    EPSC112-01A/LP
    112-01A/LP
    1000 ml
    95,25 mm
    215,9 mm
    Amber
    38 - 430 mm
    1 * 12 SZT
    1 * 12 SZT - 3 750 zł
    3 750 zł
    1 * 12 SZT
       3 750 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Glass jug with screw cap with PTFE/silicone liner
    Glass jug with screw cap with PTFE/silicone liner
    EPSC111-04A/LP
    111-04A/LP
    4000 ml
    158,75 mm
    336,55 mm
    Amber
    38 - 430 mm
    1 * 4 SZT
    1 * 4 SZT - 1 800 zł
    1 800 zł
    1 * 4 SZT
       1 800 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Clear
  •  
    Capacity
    Ext.Ø
    Height
    Colour
    Cap Size
  • Boston round bottle with closed top with screw cap with PTFE/silicone liner
    Boston round bottle with closed top with screw cap with PTFE/silicone liner
    EPSC114-125CT/LP
    114-125CT/LP
    125 ml
    47,63 mm
    111,13 mm
    Clear
    24 - 414 mm
    1 * 12 SZT
    1 * 12 SZT - 1 410 zł
    1 410 zł
    1 * 12 SZT
       1 410 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Boston round bottle with closed top with screw cap with PTFE/silicone liner
    Boston round bottle with closed top with screw cap with PTFE/silicone liner
    EPSC114-250C/LP
    114-250C/LP
    250 ml
    60,33 mm
    136,53 mm
    Clear
    24 - 414 mm
    1 * 24 SZT
    1 * 24 SZT - 2 800 zł
    2 800 zł
    1 * 24 SZT
       2 800 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Glass vial with septa closure, particulate cleaned with PTFE/silicone liner
    Glass vial with septa closure, particulate cleaned with PTFE/silicone liner
    EPSC140-40C/LP
    140-40C/LP
    40 ml
    28,58 mm
    95,25 mm
    Clear
    24 - 414 mm
    1 * 90 SZT
    1 * 90 SZT - 3 910 zł
    3 910 zł
    1 * 90 SZT
       3 910 zł
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact