Drukuj

Anti-CD9 Mouse Polyclonal Antibody [clone: eBioSN4 (SN4 C3-3A2)]

Dostawca: OriGene
ORIGTA320303EA 1610 PLN
ORIGTA320303
Anti-CD9 Mouse Polyclonal Antibody [clone: eBioSN4 (SN4 C3-3A2)]
Przeciwciała
Type: Primary
Antigen: CD9
Clonality: Polyclonal
Clone: eBioSN4 (SN4 C3-3A2)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR