Drukuj

Anti-LPIN1 Mouse Monoclonal Antibody [clone: 4C11]

Dostawca: OriGene
ORIGCF806417EA 3950 PLN
ORIGCF806417
Anti-LPIN1 Mouse Monoclonal Antibody [clone: 4C11]
Przeciwciała
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: LPIN1
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR